Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153634.2 | NP_705898.1 | 20 | 2902 | ||
XM_017018851.1 | XP_016874340.1 | 21 | 3875 | |||
XM_017018852.1 | XP_016874341.1 | 20 | 2848 | |||
GenBank mRNA | AK074223.1 | - | 6 | 1765 | ||
AK094998.1 | - | 8 | 2488 | |||
BC046366.1 | - | 8 | 2218 | |||
BX538055.1 | - | 20 | 2929 |