Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318316.1 | NP_001305245.1 | 4 | 302 | ||
NM_172002.4 | NP_741999.3 | 4 | 548 | |||
NR_134560.1 | 4 | 493 | ||||
XM_005261359.3 | XP_005261416.1 | 4 | 549 | |||
XM_017028619.1 | XP_016884108.1 | 4 | 519 | |||
XM_017028620.1 | XP_016884109.1 | 4 | 352 | |||
GenBank mRNA | AY191719.1 | - | 4 | 498 | ||
BC000004.1 | - | 4 | 557 | |||
BC065569.1 | - | 4 | 521 | 6171959 | ||
BG696148.1 | - | 4 | 482 | |||
CR456462.1 | - | 4 | 516 | |||
HY057929.1 | - | 4 | 302 |