Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199777.1 | NP_001186706.1 | 2 | 536 | ||
NM_172240.2 | NP_758440.1 | 3 | 555 | |||
GenBank mRNA | AK074772.1 | - | 3 | 293 | ||
AK123106.1 | - | 2 | 533 | |||
AL832918.1 | - | 1 | 20 | |||
BC026080.2 | - | 3 | 317 | 4827689 | ||
DA851891.1 | - | 2 | 536 | |||
EF011619.1 | - | 3 | 317 |