Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBB2 SIRNA Assays ›
Gene Symbol:
APBB2
Gene Name:
amyloid beta precursor protein binding family B member 2
Gene Aliases:
FE65L, FE65L1
Chromosome Location:
Chr.4: 40810027 - 41216714 on Build GRCh38
UniGene ID:
Hs.479602
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
40565
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166050.1
NP_001159522.1
14
2330
NM_001166051.1
NP_001159523.1
4
351
NM_001166052.1
NP_001159524.1
3
345
NM_001166053.1
NP_001159525.1
4
347
NM_001166054.1
NP_001159526.1
3
341
NM_004307.1
NP_004298.1
14
2333
NM_173075.4
NP_775098.2
13
2267
XM_005248101.3
XP_005248158.1
13
2264
XM_006714005.3
XP_006714068.1
15
2336
XM_006714007.3
XP_006714070.1
15
2334
XM_006714008.3
XP_006714071.1
14
2238
XM_006714009.3
XP_006714072.1
14
2236
XM_006714010.3
XP_006714073.1
15
2333
XM_006714011.3
XP_006714074.1
14
2273
XM_006714012.3
XP_006714075.1
14
2270
XM_011513686.2
XP_011511988.1
16
2358
XM_011513687.2
XP_011511989.1
13
2267
XM_017008142.1
XP_016863631.1
15
2257
XM_017008143.1
XP_016863632.1
16
2355
XM_017008144.1
XP_016863633.1
14
2235
XM_017008145.1
XP_016863634.1
13
2232
XM_017008146.1
XP_016863635.1
14
2270
XM_017008147.1
XP_016863636.1
14
2265
XM_017008148.1
XP_016863637.1
13
2169
XM_017008149.1
XP_016863638.1
14
2267
XM_017008150.1
XP_016863639.1
14
2194
XM_017008151.1
XP_016863640.1
13
2175
XM_017008152.1
XP_016863641.1
13
2172
XM_017008153.1
XP_016863642.1
13
2188
XM_017008154.1
XP_016863643.1
12
2169
XM_017008155.1
XP_016863644.1
12
2166
XM_017008156.1
XP_016863645.1
13
2207
XM_017008157.1
XP_016863646.1
12
2106
XM_017008158.1
XP_016863647.1
12
2201
GenBank mRNA
AK226179.1
-
14
2247
AK289838.1
-
7
817
AK299800.1
-
3
316
AK304247.1
-
14
2201
BC027946.1
-
13
2107
5203925
BC088372.1
-
5
521
BU184203.1
-
4
343
BX440903.2
-
4
341
CX782504.1
-
3
335
DC372060.1
-
3
341
DC377357.1
-
3
304
DC377693.1
-
4
333
DC400218.1
-
3
345
KU177906.1
-
4
416
KU177907.1
-
5
500
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
regulation of transcription, DNA-templated
cell cycle arrest
axon guidance
extracellular matrix organization
negative regulation of cell growth
intracellular signal transduction
positive regulation of apoptotic process
negative regulation of apoptotic process
beta-amyloid binding
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0