Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLITRK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184749.2 | NP_001171678.1 | 2 | 566 | ||
NM_001184750.2 | NP_001171679.1 | 2 | 369 | |||
NM_173078.4 | NP_775101.1 | 2 | 519 | |||
XM_005262365.4 | XP_005262422.1 | 2 | 319 | |||
XM_011531265.2 | XP_011529567.1 | 2 | 364 | |||
GenBank mRNA | AK092016.1 | - | 2 | 236 | ||
AK315044.1 | - | 2 | 369 | |||
BC040986.1 | - | 2 | 528 | 4811695 |