Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNA2D4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172364.4 | NP_758952.4 | 38 | 4371 | ||
GenBank mRNA | AF516695.1 | - | 38 | 4503 | ||
AL137658.1 | - | 15 | 1910 | |||
BC048288.1 | - | 10 | 1477 | |||
BC150186.1 | - | 37 | 4294 | 40148321 | ||
BX537436.1 | - | 36 | 4971 | |||
BX537437.1 | - | 38 | 4372 |