Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APBB2 SIRNA Assays ›
Gene Symbol:
APBB2
Gene Name:
amyloid beta precursor protein binding family B member 2
Gene Aliases:
FE65L, FE65L1
Chromosome Location:
Chr.4: 40810027 - 41216714 on Build GRCh38
UniGene ID:
Hs.479602
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
40743
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APBB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166050.1
NP_001159522.1
18
4305
NM_001166051.1
NP_001159523.1
8
2326
NM_001166052.1
NP_001159524.1
7
2320
NM_001166053.1
NP_001159525.1
8
2322
NM_001166054.1
NP_001159526.1
7
2316
NM_004307.1
NP_004298.1
18
4308
NM_173075.4
NP_775098.2
17
4239
XM_005248101.3
XP_005248158.1
17
4239
XM_006714005.3
XP_006714068.1
19
4311
XM_006714007.3
XP_006714070.1
19
4309
XM_006714008.3
XP_006714071.1
18
4213
XM_006714009.3
XP_006714072.1
18
4211
XM_006714010.3
XP_006714073.1
19
4308
XM_006714011.3
XP_006714074.1
18
4248
XM_006714012.3
XP_006714075.1
18
4245
XM_011513686.2
XP_011511988.1
20
4333
XM_011513687.2
XP_011511989.1
17
4242
XM_017008142.1
XP_016863631.1
19
4232
XM_017008143.1
XP_016863632.1
20
4330
XM_017008144.1
XP_016863633.1
18
4210
XM_017008145.1
XP_016863634.1
17
4207
XM_017008146.1
XP_016863635.1
18
4245
XM_017008147.1
XP_016863636.1
18
4240
XM_017008148.1
XP_016863637.1
17
4144
XM_017008149.1
XP_016863638.1
18
4242
XM_017008150.1
XP_016863639.1
18
4169
XM_017008151.1
XP_016863640.1
17
4150
XM_017008152.1
XP_016863641.1
17
4147
XM_017008153.1
XP_016863642.1
17
4163
XM_017008154.1
XP_016863643.1
16
4144
XM_017008155.1
XP_016863644.1
16
4141
XM_017008156.1
XP_016863645.1
17
4182
XM_017008157.1
XP_016863646.1
16
4081
XM_017008158.1
XP_016863647.1
16
4176
GenBank mRNA
BC027946.1
-
17
4079
5203925
BM546260.1
-
1
757
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron migration
regulation of transcription, DNA-templated
cell cycle arrest
axon guidance
extracellular matrix organization
negative regulation of cell growth
intracellular signal transduction
positive regulation of apoptotic process
negative regulation of apoptotic process
beta-amyloid binding
protein binding
transcription factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0