Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCMDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136160.1 | NP_001129632.1 | 7 | 780 | ||
NM_001136161.1 | NP_001129633.1 | 7 | 815 | |||
NM_173518.4 | NP_775789.3 | 7 | 815 | |||
XM_005251174.2 | XP_005251231.1 | 6 | 677 | |||
XM_006716427.3 | XP_006716490.1 | 7 | 753 | |||
XM_006716429.2 | XP_006716492.1 | 7 | 774 | |||
XM_006716433.3 | XP_006716496.1 | 5 | 495 | |||
XM_011517467.2 | XP_011515769.1 | 7 | 774 | |||
XM_011517468.2 | XP_011515770.1 | 7 | 774 | |||
XM_011517469.2 | XP_011515771.1 | 7 | 774 | |||
XM_017013140.1 | XP_016868629.1 | 5 | 495 | |||
GenBank mRNA | AK098558.1 | - | 3 | 351 | ||
AK302169.1 | - | 7 | 780 | |||
AK302672.1 | - | 7 | 774 | |||
AK303003.1 | - | 5 | 571 | |||
AK304170.1 | - | 6 | 677 | |||
BC034576.1 | - | 7 | 815 | 4825975 | ||
BC063108.1 | - | 6 | 693 | 4690709 | ||
BX537587.1 | - | 7 | 775 |