Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199085.1 | NP_001186014.1 | 9 | 2004 | ||
NM_001199089.1 | NP_001186018.1 | 9 | 1717 | |||
NM_001199091.1 | NP_001186020.1 | 9 | 1845 | |||
NM_001199092.1 | NP_001186021.1 | 6 | 689 | |||
NM_173533.3 | NP_775804.2 | 9 | 2004 | |||
XM_005244934.1 | XP_005244991.1 | 9 | 1789 | |||
XM_005244935.4 | XP_005244992.1 | 9 | 1694 | |||
XM_017000473.1 | XP_016855962.1 | 9 | 1710 | |||
GenBank mRNA | AK092142.1 | - | 6 | 702 | ||
AK126889.1 | - | 9 | 1845 | |||
AK127370.1 | - | 9 | 1736 | |||
BC065707.1 | - | 6 | 813 | |||
BC089387.1 | - | 7 | 906 | |||
BC130532.1 | - | 9 | 1681 | 40146563 | ||
BC144058.1 | - | 9 | 1736 | 9052578 |