Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242702.1 | NP_001229631.1 | 3, 4 | 578 | ||
NM_173576.2 | NP_775847.2 | 3, 4 | 581 | |||
XM_017016105.1 | XP_016871594.1 | 3, 4 | 618 | |||
XM_017016106.1 | XP_016871595.1 | 3, 4 | 809 | |||
XM_017016107.1 | XP_016871596.1 | 2, 3 | 486 | |||
XM_017016108.1 | XP_016871597.1 | 2, 3 | 486 | |||
GenBank mRNA | AK125339.1 | - | 3, 4 | 583 | ||
BC036207.1 | - | 3, 4 | 547 | 5262515 | ||
DB285243.1 | - | 2, 3 | 486 |