Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173528.2 | NP_775799.2 | 4 | 495 | ||
XM_006720408.2 | XP_006720471.1 | 5 | 648 | |||
XM_011521280.1 | XP_011519582.1 | 4 | 489 | |||
XM_017021963.1 | XP_016877452.1 | 5 | 573 | |||
GenBank mRNA | AK095934.1 | - | 5 | 648 | ||
AK302120.1 | - | 4 | 533 | |||
AK307565.1 | - | 4 | 490 | |||
BC062471.1 | - | 4 | 495 | 30390735 |