Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173545.2 | NP_775816.1 | 5 | 726 | ||
GenBank mRNA | AK290841.1 | - | 5 | 726 | ||
AK308186.1 | - | 4 | 616 | |||
BC041144.1 | - | 5 | 675 | 6042653 |