Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173566.2 | NP_775837.2 | 5 | 5989 | ||
XM_006724219.3 | XP_006724282.1 | 5 | 6549 | |||
XM_006724220.3 | XP_006724283.1 | 5 | 5951 | |||
XM_006724221.2 | XP_006724284.1 | 4 | 5505 | |||
XM_011530080.2 | XP_011528382.1 | 6 | 6757 | |||
XM_011530081.2 | XP_011528383.1 | 5 | 6633 | |||
XM_017028736.1 | XP_016884225.1 | 5 | 7709 | |||
XM_017028737.1 | XP_016884226.1 | 4 | 7147 | |||
GenBank mRNA | AK130944.1 | - | 2 | 1803 | ||
BC040859.1 | - | 2 | 703 | 5535580 | ||
BX647272.1 | - | 2 | 292 | |||
CT841517.1 | - | 4 | 5821 |