Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199085.1 | NP_001186014.1 | 18 | 3706 | ||
NM_001199089.1 | NP_001186018.1 | 18 | 3419 | |||
NM_001199091.1 | NP_001186020.1 | 17 | 3385 | |||
NM_001199092.1 | NP_001186021.1 | 14 | 2229 | |||
NM_173533.3 | NP_775804.2 | 17 | 3544 | |||
XM_005244934.1 | XP_005244991.1 | 18 | 3491 | |||
XM_005244935.4 | XP_005244992.1 | 18 | 3396 | |||
XM_017000473.1 | XP_016855962.1 | 17 | 3250 | |||
GenBank mRNA | AK092142.1 | - | 14 | 2241 | ||
AK126889.1 | - | 17 | 3385 | |||
AK127370.1 | - | 17 | 3276 | |||
BC043368.1 | - | 9 | 1556 | |||
BC089387.1 | - | 15 | 2446 |