Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR14L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173566.2 | NP_775837.2 | 7, 8 | 6308 | ||
XM_006724219.3 | XP_006724282.1 | 7, 8 | 6868 | |||
XM_006724220.3 | XP_006724283.1 | 7, 8 | 6270 | |||
XM_006724221.2 | XP_006724284.1 | 6, 7 | 5824 | |||
XM_011530080.2 | XP_011528382.1 | 8, 9 | 7076 | |||
XM_011530081.2 | XP_011528383.1 | 7, 8 | 6952 | |||
XM_017028736.1 | XP_016884225.1 | 7, 8 | 8028 | |||
XM_017028737.1 | XP_016884226.1 | 6, 7 | 7466 | |||
GenBank mRNA | AK123082.1 | - | 2, 3 | 284 | ||
BC040859.1 | - | 4, 5 | 1022 | 5535580 | ||
BX647272.1 | - | 4, 5 | 611 | |||
CT841517.1 | - | 6, 7 | 6140 |