Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001200049.2 | NP_001186978.2 | 4, 5 | 480 | ||
GenBank mRNA | AK098820.1 | - | 4, 5 | 455 | ||
BC044661.1 | - | 4, 5 | 438 | |||
BC146900.1 | - | 3, 4 | 263 | 9056828 | ||
BC146908.1 | - | 3, 4 | 263 | 9056836 | ||
BX376477.2 | - | 4, 5 | 480 | |||
EL734348.1 | - | 4, 5 | 379 |