Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000397.3 | NP_000388.2 | 13 | 3110 | ||
GenBank mRNA | BC032720.1 | - | 13 | 3096 | 5505170 | |
X04011.1 | - | 13 | 3064 |