Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173598.4 | NP_775869.3 | 6, 7 | 1217 | ||
XM_011538224.2 | XP_011536526.1 | 6, 7 | 1552 | |||
XM_011538225.2 | XP_011536527.1 | 6, 7 | 2011 | |||
XM_011538226.2 | XP_011536528.1 | 6, 7 | 1552 | |||
XM_011538228.2 | XP_011536530.1 | 4, 5 | 376 | |||
XM_011538229.2 | XP_011536531.1 | 6, 7 | 1552 | |||
XM_017019208.1 | XP_016874697.1 | 6, 7 | 1552 | |||
XM_017019209.1 | XP_016874698.1 | 6, 7 | 1552 | |||
GenBank mRNA | AK098831.1 | - | 4, 5 | 395 | ||
AY345972.1 | - | 4, 5 | 886 | |||
BC107106.1 | - | 4, 5 | 881 | |||
BC107107.1 | - | 4, 5 | 881 | |||
BC127603.1 | - | 4, 5 | 886 | 40019272 |