Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC9A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173653.3 | NP_775924.1 | 3 | 656 | ||
XM_011512704.2 | XP_011511006.1 | 3 | 629 | |||
XM_017006202.1 | XP_016861691.1 | 3 | 629 | |||
XM_017006203.1 | XP_016861692.1 | 2 | 414 | |||
GenBank mRNA | AB089794.1 | - | 3 | 447 | ||
AK301816.1 | - | 2 | 382 | |||
AK315107.1 | - | 3 | 583 | |||
AL832304.1 | - | 3 | 521 | |||
AY254100.1 | - | 3 | 571 | |||
BC035779.1 | - | 3 | 572 | 5590356 |