Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080448.2 | NP_001073917.2 | 8 | 1969 | ||
NM_001278300.1 | NP_001265229.1 | 5 | 381 | |||
NM_173655.3 | NP_775926.1 | 6 | 497 | |||
XM_006713592.3 | XP_006713655.1 | 9 | 2017 | |||
XM_017006210.1 | XP_016861699.1 | 9 | 2017 | |||
XM_017006211.1 | XP_016861700.1 | 10 | 2042 | |||
XM_017006212.1 | XP_016861701.1 | 9 | 1711 | |||
XM_017006213.1 | XP_016861702.1 | 9 | 2069 | |||
XM_017006214.1 | XP_016861703.1 | 8 | 1337 | |||
XM_017006215.1 | XP_016861704.1 | 4 | 366 | |||
XM_017006216.1 | XP_016861705.1 | 3 | 316 | |||
XM_017006217.1 | XP_016861706.1 | 5 | 413 | |||
XM_017006218.1 | XP_016861707.1 | 3 | 216 | |||
XM_017006220.1 | XP_016861709.1 | 5 | 381 | |||
XM_017006221.1 | XP_016861710.1 | 3 | 327 | |||
XM_017006222.1 | XP_016861711.1 | 4 | 375 | |||
XM_017006223.1 | XP_016861712.1 | 5 | 381 | |||
GenBank mRNA | AK302040.1 | - | 5 | 364 | ||
AK302080.1 | - | 4 | 271 | |||
AK310249.1 | - | 4 | 300 | |||
AL133666.1 | - | 5 | 444 | |||
DC398430.1 | - | 4 | 319 | |||
DN991634.1 | - | 4 | 432 |