Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173687.3 | NP_775958.1 | 6 | 900 | ||
NR_126398.1 | 8 | 1082 | ||||
NR_126399.1 | 6 | 891 | ||||
GenBank mRNA | AK094450.1 | - | 6 | 848 | ||
BC063497.1 | - | 6 | 802 | |||
BC073830.1 | - | 8 | 1030 | |||
BC120993.2 | - | 6 | 826 |