Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031748.3 | NP_001026918.2 | 6 | 569 | ||
NM_001319247.1 | NP_001306176.1 | 6 | 569 | |||
NR_135051.1 | 5 | 430 | ||||
XM_005268806.3 | XP_005268863.1 | 5 | 379 | |||
XM_011538231.2 | XP_011536533.1 | 5 | 606 | |||
GenBank mRNA | AK097445.1 | - | 5 | 379 | ||
AL705293.1 | - | 6 | 545 | |||
BC038754.1 | - | 6 | 566 | 5269851 | ||
BC048120.1 | - | 6 | 553 | 5266211 | ||
BI461967.1 | - | 6 | 570 | |||
EU668334.1 | - | 5 | 379 | |||
GQ472216.1 | - | 5 | 379 |