Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000774.4 | NP_000765.2 | 10 | 1739 | ||
NR_135528.1 | 10 | 1681 | ||||
XM_011526553.2 | XP_011524855.1 | 7 | 1740 | |||
XM_011526555.2 | XP_011524857.1 | 7 | 1682 | |||
XM_017026385.1 | XP_016881874.1 | 9 | 2070 | |||
GenBank mRNA | J02906.1 | - | 10 | 1722 |