Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000966.5 | NP_000957.1 | 10 | 2693 | ||
NM_001042728.2 | NP_001036193.1 | 8 | 2481 | |||
NM_001243730.1 | NP_001230659.1 | 9 | 2367 | |||
NM_001243731.1 | NP_001230660.1 | 8 | 2218 | |||
NM_001243732.1 | NP_001230661.1 | 7 | 2332 | |||
GenBank mRNA | M38258.1 | - | 8 | 2347 |