Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFAND4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128324.2 | NP_001121796.1 | 2, 3 | 444 | ||
NM_174890.3 | NP_777550.2 | 2, 3 | 792 | |||
NR_104256.1 | 2, 3 | 444 | ||||
XM_017016932.1 | XP_016872421.1 | 2, 3 | 536 | |||
XM_017016933.1 | XP_016872422.1 | 2, 3 | 403 | |||
XM_017016934.1 | XP_016872423.1 | 2, 3 | 456 | |||
XM_017016935.1 | XP_016872424.1 | 1, 2 | 206 | |||
XM_017016946.1 | XP_016872435.1 | 2, 3 | 403 | |||
GenBank mRNA | AF311324.1 | - | 2, 3 | 408 | ||
AK090704.1 | - | 3, 4 | 618 | |||
AK097924.1 | - | 2, 3 | 411 | |||
AK292469.1 | - | 2, 3 | 393 | |||
AK314395.1 | - | 2, 3 | 404 | |||
BC045587.1 | - | 2, 3 | 444 | 5269483 |