Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175068.2 | NP_778238.1 | 7, 8 | 1374 | ||
XM_011538257.2 | XP_011536559.1 | 6, 7 | 2852 | |||
GenBank mRNA | AJ508776.1 | - | 7, 8 | 1374 | ||
BC109212.2 | - | 6, 7 | 1732 | 40007200 |