Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APTX SIRNA Assays ›
Gene Symbol:
APTX
Gene Name:
aprataxin
Gene Aliases:
AOA, AOA1, AXA1, EAOH, EOAHA, FHA-HIT
Chromosome Location:
Chr.9: 32883872 - 33025129 on Build GRCh38
UniGene ID:
Hs.20158
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
41422
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: APTX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001195248.1
NP_001182177.1
3
246
NM_001195249.1
NP_001182178.1
3
415
NM_001195251.1
NP_001182180.1
4
352
NM_001195252.1
NP_001182181.1
3
246
NM_175069.2
NP_778239.1
3
246
NM_175073.2
NP_778243.1
4
352
NR_036576.1
3
241
NR_036577.1
3
246
NR_036578.1
3
263
NR_036579.1
3
410
XM_006716791.3
XP_006716854.1
3
254
XM_006716792.2
XP_006716855.1
3
272
XM_017014831.1
XP_016870320.1
3
246
XM_017014832.1
XP_016870321.1
3
246
XM_017014833.1
XP_016870322.1
3
246
XM_017014834.1
XP_016870323.1
3
246
XM_017014835.1
XP_016870324.1
3
246
XM_017014836.1
XP_016870325.1
3
246
XM_017014838.1
XP_016870327.1
3
228
GenBank mRNA
AB603741.1
-
2
142
AJ565850.1
-
3
219
AJ565851.1
-
3
224
AJ565852.1
-
3
219
AJ565853.1
-
2
222
AJ565854.1
-
2
227
AJ575566.1
-
2
222
AK000164.1
-
2
364
AK055672.1
-
3
233
AK131046.1
-
3
198
AY040777.1
-
3
221
AY208829.1
-
3
221
AY208831.1
-
3
212
AY208834.1
-
3
217
AY208835.1
-
3
201
AY208836.1
-
3
361
AY208837.1
-
4
318
AY208838.1
-
2
292
AY208839.1
-
3
355
AY208840.1
-
3
218
AY208841.1
-
2
170
AY302067.1
-
2
364
AY302068.1
-
2
359
AY302069.1
-
2
364
AY302070.1
-
2
359
AY302071.1
-
2
364
AY302072.1
-
2
359
AY302073.1
-
2
364
AY302074.1
-
2
359
BC001628.1
-
2
184
BC032650.1
-
3
287
5505120
BC068107.1
-
4
327
BC104881.1
-
3
260
8143884
BQ957536.1
-
4
335
BX538161.1
-
4
331
DA664623.1
-
2
273
Back To Top
More Information
Panther Classification:
Molecular Function -
damaged DNA-binding protein
Gene Ontology Categories:
Function(s)
Process(es)
single strand break repair
DNA ligation
cellular response to DNA damage stimulus
dephosphorylation
regulation of protein stability
response to hydrogen peroxide
nucleic acid phosphodiester bond hydrolysis
chromatin binding
damaged DNA binding
double-stranded DNA binding
double-stranded RNA binding
protein binding
phosphoglycolate phosphatase activity
DNA 5'-adenosine monophosphate hydrolase activity
metal ion binding
protein N-terminus binding
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0