Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC30A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172811.1 | NP_001166282.1 | 7 | 799 | ||
NM_001172813.1 | NP_001166284.1 | 8 | 1044 | |||
NM_001172814.1 | NP_001166285.1 | 6 | 886 | |||
NM_001172815.2 | NP_001166286.1 | 8 | 913 | |||
NM_173851.2 | NP_776250.2 | 5 | 856 | |||
GenBank mRNA | AK298294.1 | - | 9 | 1047 | ||
AL713790.1 | - | 8 | 885 | |||
AY117411.1 | - | 5 | 626 | |||
BC126446.1 | - | 6 | 725 | |||
BC143953.1 | - | 6 | 726 | |||
EF560713.1 | - | 7 | 799 |