Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257378.1 | NP_001244307.1 | 7, 8 | 737 | ||
NM_001257379.1 | NP_001244308.1 | 6, 7 | 567 | |||
NM_001257382.1 | NP_001244311.1 | 6, 7 | 637 | |||
NM_174975.4 | NP_777635.1 | 5, 6 | 519 | |||
XM_011530128.2 | XP_011528430.1 | 5, 6 | 487 | |||
GenBank mRNA | AK131358.1 | - | 6, 7 | 541 | ||
AK298079.1 | - | 6, 8 | 511 | |||
AY158086.1 | - | 5, 6 | 430 | |||
AY240872.1 | - | 5, 6 | 519 | |||
BC069641.1 | - | 5, 6 | 442 | 7262444 | ||
BC101001.2 | - | 7, 8 | 676 | 40006372 | ||
BC101002.1 | - | 6, 7 | 506 | 40006375 | ||
BC101003.1 | - | 6, 7 | 576 | 40006376 | ||
BC101004.1 | - | 5, 6 | 458 | 40006377 |