Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174978.2 | NP_777638.2 | 4 | 328 | ||
XM_011536690.2 | XP_011534992.1 | 5 | 458 | |||
XM_011536703.2 | XP_011535005.1 | 5 | 458 | |||
XM_011536704.1 | XP_011535006.1 | 5 | 459 | |||
XM_017021247.1 | XP_016876736.1 | 5 | 459 | |||
XM_017021248.1 | XP_016876737.1 | 3 | 291 | |||
GenBank mRNA | AK098187.1 | - | 4 | 313 | ||
AL832219.1 | - | 4 | 378 | |||
BC125070.1 | - | 4 | 299 | 40117603 | ||
DB447488.2 | - | 4 | 347 |