Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001508.2 | NP_001499.1 | 1, 2 | 1334 | ||
GenBank mRNA | AF034633.1 | - | 1, 2 | 865 | ||
AK122643.1 | - | 1, 2 | 1334 | |||
AK314895.1 | - | 1, 2 | 1091 | |||
BC125046.1 | - | 1, 2 | 865 | 40116106 | ||
BC125047.1 | - | 1, 2 | 854 | |||
FJ348258.1 | - | 1, 2 | 865 |
Set Membership: |
G-protein coupled receptor |