Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174918.2 | NP_777578.2 | 5 | 485 | ||
GenBank mRNA | AF461155.1 | - | 5 | 914 | ||
AF461156.1 | - | 5 | 464 | |||
BC035847.1 | - | 5 | 427 | |||
BC104796.1 | - | 5 | 485 | 8143799 | ||
BC104798.1 | - | 5 | 485 | 8143801 |