Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPN1SW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001708.2 | NP_001699.1 | 5 | 1056 |
Set Membership: |
G-protein coupled receptor |