Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC002463.1 | - | 8 | 1338 | 3345005 | |
BC008457.1 | - | 9 | 1362 | 4249847 | ||
BC010542.1 | - | 8 | 1332 | |||
D12471.1 | - | 8 | 1333 | |||
M21963.1 | - | 7 | 1210 | |||
M24096.1 | - | 7 | 1184 | |||
M24097.1 | - | 8 | 1360 | |||
M26429.1 | - | 8 | 1333 | |||
M26430.1 | - | 8 | 1335 | |||
M26431.1 | - | 8 | 1335 | |||
M26712.1 | - | 7 | 1277 | |||
M28160.1 | - | 8 | 1337 | |||
M28206.1 | - | 7 | 1273 | |||
X58536.1 | - | 9 | 1349 |