Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080770.1 | NP_001074239.1 | 3 | 275 | ||
NM_001080772.1 | NP_001074241.1 | 3 | 275 | |||
NM_002255.5 | NP_002246.5 | 3 | 275 | |||
GenBank mRNA | AF002979.1 | - | 3 | 235 | ||
AF002980.1 | - | 3 | 247 | |||
AF002981.1 | - | 3 | 247 | |||
AF002982.1 | - | 3 | 247 | |||
AF034771.1 | - | 3 | 241 | |||
AF034772.1 | - | 3 | 241 | |||
AF034773.1 | - | 3 | 241 | |||
AF276292.1 | - | 3 | 235 | |||
AF285436.1 | - | 3 | 262 | |||
AK309531.1 | - | 4 | 511 | |||
AK310066.1 | - | 3 | 517 | |||
AY223512.2 | - | 1 | 162 | |||
AY223513.1 | - | 1 | 162 | |||
AY223514.1 | - | 1 | 162 | |||
AY223515.1 | - | 1 | 162 | |||
AY250088.1 | - | 1 | 162 | |||
AY359817.1 | - | 3 | 246 | |||
AY601812.1 | - | 3 | 229 | |||
AY789058.1 | - | 3 | 229 | |||
AY789059.1 | - | 3 | 229 | |||
AY789060.1 | - | 3 | 229 | |||
AY789061.1 | - | 3 | 229 | |||
AY789062.1 | - | 3 | 229 | |||
BC041611.1 | - | 3 | 276 | 5485491 | ||
BM918119.1 | - | 275 | ||||
DA590865.1 | - | 4 | 511 | |||
DA934665.1 | - | 3 | 517 | |||
DQ266438.1 | - | 3 | 238 | |||
EU194340.1 | - | 3 | 235 | |||
EU194342.1 | - | 3 | 235 | |||
U71199.1 | - | 3 | 247 | |||
U73394.1 | - | 3 | 247 | |||
X97229.1 | - | 3 | 240 | |||
X99479.1 | - | 3 | 235 | |||
X99480.1 | - | 3 | 246 | |||
Y13054.1 | - | 3 | 235 |