Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GBA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000157.3 | NP_000148.2 | 4 | 505 | ||
NM_001005741.2 | NP_001005741.1 | 5 | 764 | |||
NM_001005742.2 | NP_001005742.1 | 5 | 745 | |||
NM_001171811.1 | NP_001165282.1 | 3 | 581 | |||
GenBank mRNA | AK291911.1 | - | 5 | 743 | ||
AK300829.1 | - | 3 | 418 | |||
AK300876.1 | - | 5 | 745 | |||
AK302000.1 | - | 3 | 330 | |||
AK311242.1 | - | 5 | 353 | |||
AK312502.1 | - | 5 | 561 | |||
AL547558.3 | - | 6 | 676 | |||
AU122729.1 | - | 6 | 665 | |||
AU131071.1 | - | 5 | 573 | |||
AU140959.1 | - | 4 | 489 | |||
AU141794.1 | - | 5 | 580 | |||
BC003356.1 | - | 4 | 444 | 2899915 | ||
BI458641.1 | - | 5 | 706 | |||
BX382279.2 | - | 6 | 663 | |||
D13286.1 | - | 4 | 444 | |||
DC297079.1 | - | 4 | 505 | |||
K02920.1 | - | 3 | 324 | |||
KJ690771.1 | - | 4 | 388 | |||
KJ690772.1 | - | 4 | 388 | |||
KJ690773.1 | - | 4 | 388 | |||
M16328.1 | - | 4 | 774 | |||
M19285.1 | - | 4 | 461 |