Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000331.5 | NP_000322.2 | 4 | 483 | ||
NM_001178006.2 | NP_001171477.1 | 5 | 397 | |||
NM_199161.4 | NP_954630.1 | 4 | 336 | |||
GenBank mRNA | BC007022.1 | - | 4 | 297 | 4071612 | |
BC105796.1 | - | 5 | 362 | 4722684 | ||
BG533276.1 | - | 445 | ||||
BG562565.1 | - | 4 | 336 | |||
BG565078.1 | - | 4 | 445 | |||
BG567902.1 | - | 388 | ||||
BQ691948.1 | - | 3 | 199 | |||
BX102362.1 | - | 4 | 403 | |||
CD102084.1 | - | 4 | 300 | |||
CR542241.1 | - | 3 | 260 | |||
M10906.1 | - | 3 | 260 | |||
M23698.1 | - | 3 | 260 | |||
X51439.1 | - | 1 | 46 | |||
X51441.1 | - | 1 | 46 |
Gene Symbol: SAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X51442.1 | - | 1 | 46 |