Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTSB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317237.1 | NP_001304166.1 | 10 | 1942 | ||
NM_001908.4 | NP_001899.1 | 10 | 1973 | |||
NM_147780.3 | NP_680090.1 | 12 | 2135 | |||
NM_147781.3 | NP_680091.1 | 11 | 2092 | |||
NM_147782.3 | NP_680092.1 | 11 | 2061 | |||
NM_147783.3 | NP_680093.1 | 11 | 2047 | |||
XM_006716244.2 | XP_006716307.1 | 12 | 2159 | |||
XM_006716245.2 | XP_006716308.1 | 10 | 2263 | |||
XM_011543812.2 | XP_011542114.1 | 12 | 2068 | |||
XM_017013097.1 | XP_016868586.1 | 11 | 2049 | |||
XM_017013098.1 | XP_016868587.1 | 11 | 2049 | |||
XM_017013099.1 | XP_016868588.1 | 12 | 2091 | |||
XM_017013100.1 | XP_016868589.1 | 11 | 1993 | |||
XM_017013101.1 | XP_016868590.1 | 7 | 1516 | |||
GenBank mRNA | AK075393.1 | - | 10 | 1949 | ||
AK092070.1 | - | 11 | 1982 | |||
AK097384.1 | - | 5 | 1901 | |||
AK128472.1 | - | 1 | 2564 | |||
AK130184.1 | - | 10 | 1910 | |||
BC010240.1 | - | 11 | 1990 | 4154307 | ||
BC095408.1 | - | 10 | 1910 | 30334082 | ||
BX647765.1 | - | 10 | 1833 | |||
BX647952.1 | - | 10 | 1913 | |||
BX648239.1 | - | 5 | 7684 | |||
L16510.1 | - | 11 | 1968 | |||
M14221.1 | - | 11 | 1975 |