Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003329.3 | NP_003320.2 | 3 | 357 | ||
GenBank mRNA | AF085844.1 | - | 3 | 164 | ||
AF276919.1 | - | 2 | 152 | |||
AF313911.1 | - | 3 | 162 | |||
AK289508.1 | - | 3 | 278 | |||
AW160756.1 | - | 3 | 357 | |||
AY004872.1 | - | 3 | 162 | |||
BC003377.1 | - | 3 | 209 | 2901084 | ||
BC054866.1 | - | 3 | 228 | 6463280 | ||
CR407665.1 | - | 3 | 152 | |||
J04026.1 | - | 3 | 215 | |||
JQ313905.1 | - | 3 | 152 | |||
X77584.1 | - | 3 | 232 |