Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIST2H4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003548.2 | NP_003539.1 | 1 | 107 | ||
GenBank mRNA | BC019846.1 | - | 1 | 86 | ||
BC093969.1 | - | 1 | 101 | 7939814 | ||
BC108260.1 | - | 1 | 96 | |||
BC112193.1 | - | 1 | 101 | 8327661 | ||
CR542172.1 | - | 1 | 79 |
Gene Symbol: HIST2H4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034077.4 | NP_001029249.1 | 1 | 107 |