Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004542.3 | NP_004533.1 | 3 | 155 | ||
XM_017026833.1 | XP_016882322.1 | 4 | 572 | |||
GenBank mRNA | AF044955.1 | - | 3 | 147 | ||
AF070653.1 | - | 3 | 156 | |||
AK289489.1 | - | 3 | 153 | |||
BC022369.1 | - | 3 | 152 | 4737656 | ||
BC061644.1 | - | 3 | 155 | 5748578 | ||
CR542143.1 | - | 2 | 128 |