Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004680.2 | NP_004671.1 | 1 | 1390 | ||
NM_170723.1 | NP_733841.1 | 1 | 1390 | |||
XM_011531512.1 | XP_011529814.1 | 1 | 1392 | |||
GenBank mRNA | AF000981.1 | - | 1 | 1345 | ||
AF080597.1 | - | 1 | 1345 | |||
AK292375.1 | - | 1 | 1412 | |||
BC033041.1 | - | 1 | 1395 | |||
BC132955.1 | - | 1 | 1159 | 40146977 |
Gene Symbol: CDY1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003894.1 | NP_001003894.1 | 1 | 1390 | ||
NM_001003895.1 | NP_001003895.1 | 1 | 1390 | |||
XM_011531484.1 | XP_011529786.1 | 1 | 1392 | |||
GenBank mRNA | AK292318.1 | - | 1 | 1363 | ||
BC132929.1 | - | 1 | 1159 | 40146951 |