Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCL1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004918.3 | NP_004909.1 | 1, 2 | 217 | ||
GenBank mRNA | AB018563.1 | - | 1, 2 | 196 | ||
AF110466.1 | - | 1, 2 | 193 | |||
AF137027.1 | - | 1, 2 | 212 | |||
BC041616.1 | - | 1, 2 | 191 | 5816694 | ||
BC051000.1 | - | 1, 2 | 217 | 5813497 | ||
BQ058954.1 | - | 1, 2 | 80 | |||
CR457087.1 | - | 1, 2 | 166 |