Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316307.1 | NP_001303236.1 | 4, 5 | 387 | ||
NM_005052.2 | NP_005043.1 | 4, 5 | 387 | |||
GenBank mRNA | AF008591.1 | - | 4, 5 | 351 | ||
AF498966.1 | - | 4, 5 | 281 | |||
AW162931.1 | - | 4, 5 | 419 | |||
BC009605.1 | - | 4, 5 | 388 | 4284122 | ||
BC015197.1 | - | 4, 5 | 388 | 4275237 | ||
BF941478.1 | - | 4, 5 | 266 | |||
BI668056.1 | - | 4, 5 | 341 | |||
BT019443.1 | - | 4, 5 | 281 |