Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003329.3 | NP_003320.2 | 3 | 382 | ||
GenBank mRNA | AF085844.1 | - | 3 | 189 | ||
AF276919.1 | - | 2 | 177 | |||
AF313911.1 | - | 3 | 187 | |||
AK289508.1 | - | 3 | 303 | |||
AW160756.1 | - | 3 | 382 | |||
AY004872.1 | - | 3 | 187 | |||
BC003377.1 | - | 3 | 234 | 2901084 | ||
BC054866.1 | - | 3 | 253 | 6463280 | ||
CR407665.1 | - | 3 | 177 | |||
J04026.1 | - | 3 | 240 | |||
JQ313905.1 | - | 3 | 177 | |||
X77584.1 | - | 3 | 257 |