Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004146.5 | NP_004137.2 | 3 | 527 | ||
XM_011528039.2 | XP_011526341.1 | 3 | 1119 | |||
GenBank mRNA | AF112200.1 | - | 3 | 513 | ||
BC002595.2 | - | 3 | 512 | 3140806 | ||
BU565399.1 | - | 528 |