Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282382.1 | NP_001269311.1 | 2 | 630 | ||
NM_001282383.1 | NP_001269312.1 | 3 | 484 | |||
NM_004219.3 | NP_004210.1 | 3 | 323 | |||
GenBank mRNA | AF075242.1 | - | 2 | 247 | ||
AF095287.1 | - | 3 | 263 | |||
AJ223953.1 | - | 2 | 332 | |||
AK312209.1 | - | 3 | 301 | |||
BC101834.1 | - | 3 | 256 | 8069340 | ||
BC108894.1 | - | 2 | 227 | |||
BC108895.1 | - | 2 | 227 | |||
BC112071.1 | - | 3 | 256 | 8327539 | ||
BC113842.1 | - | 2 | 227 | |||
BC113896.1 | - | 2 | 227 | |||
BC128193.1 | - | 2 | 227 | 40113885 | ||
BC128194.1 | - | 2 | 227 | 40113886 | ||
BU182149.1 | - | 3 | 479 | |||
CR457135.1 | - | 2 | 227 | |||
CR541685.1 | - | 2 | 227 | |||
HY006026.1 | - | 3 | 324 |