Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002145.3 | NP_002136.1 | 1 | 242 | ||
GenBank mRNA | BC074805.2 | - | 1 | 142 | 30915528 | |
BC074806.2 | - | 1 | 142 | 30915314 | ||
X16665.1 | - | 1 | 200 |