Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005293.2 | NP_005284.2 | 2 | 362 | ||
GenBank mRNA | AB209066.1 | - | 1 | 20 | ||
BC117409.1 | - | 1 | 366 | 40125960 | ||
DR006873.1 | - | 2 | 389 | |||
JQ308168.1 | - | 1 | 273 |