Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C19orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198476.3 | NP_940878.3 | 6 | 2284 | ||
XM_005258776.4 | XP_005258833.1 | 6 | 2854 | |||
XM_005258778.4 | XP_005258835.1 | 6 | 2273 | |||
XM_011526774.2 | XP_011525076.1 | 8 | 2331 | |||
XM_011526776.2 | XP_011525078.1 | 8 | 2259 | |||
XM_011526777.2 | XP_011525079.1 | 7 | 2214 | |||
XM_011526778.2 | XP_011525080.1 | 7 | 2142 | |||
XM_011526779.2 | XP_011525081.1 | 7 | 1727 | |||
XM_011526783.2 | XP_011525085.1 | 6 | 1941 | |||
XM_011526784.2 | XP_011525086.1 | 9 | 1654 | |||
XM_011526785.2 | XP_011525087.1 | 8 | 1740 | |||
XM_017026620.1 | XP_016882109.1 | 9 | 2005 | |||
XM_017026621.1 | XP_016882110.1 | 8 | 1960 | |||
XM_017026622.1 | XP_016882111.1 | 6 | 1523 | |||
XM_017026623.1 | XP_016882112.1 | 5 | 1824 | |||
XM_017026624.1 | XP_016882113.1 | 9 | 1283 | |||
XM_017026626.1 | XP_016882115.1 | 7 | 2187 | |||
XM_017026627.1 | XP_016882116.1 | 5 | 2740 | |||
GenBank mRNA | AK123126.1 | - | 9 | 1273 | ||
BC090894.1 | - | 6 | 2050 | 30377779 |
Gene Symbol: HKR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026699.1 | XP_016882188.1 | 1 | 2826 |
Gene Symbol: LOC253805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC028622.2 | - | 1 | 2123 | ||
BC050316.1 | - | 1 | 2116 |
Gene Symbol: MDM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145337.2 | NP_001138809.1 | 11 | 2540 | ||
NM_001145339.2 | NP_001138811.1 | 9 | 2766 | |||
NM_001145340.2 | NP_001138812.1 | 5 | 2093 | |||
NM_001278462.1 | NP_001265391.1 | 6 | 2171 | |||
NM_002392.5 | NP_002383.2 | 11 | 2931 | |||
XM_005268872.4 | XP_005268929.1 | 11 | 2859 | |||
XM_006719399.3 | XP_006719462.1 | 9 | 2514 | |||
XM_006719400.3 | XP_006719463.1 | 10 | 2820 |
Gene Symbol: PITPNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK126944.1 | - | 4 | 1386 |
Gene Symbol: SNHG20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027058.1 | 3 | 1786 | |||
GenBank mRNA | AF130050.1 | - | 2 | 1411 | ||
AK098448.1 | - | 1 | 533 | |||
BX648413.1 | - | 3 | 1745 | |||
FN357117.1 | - | 3 | 1680 | |||
FN357118.1 | - | 4 | 958 | |||
FN357119.1 | - | 3 | 854 |
Gene Symbol: TDGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174136.1 | NP_001167607.1 | 6 | 956 | ||
NM_003212.3 | NP_003203.1 | 6 | 1346 | |||
GenBank mRNA | BC022393.1 | - | 6 | 1143 | 4615416 | |
BC067844.1 | - | 6 | 1145 | 5286020 | ||
CX785985.1 | - | 2 | 608 | |||
X14253.1 | - | 6 | 1210 |
Gene Symbol: ZNF506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL136548.1 | - | 6 | 3661 |